Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ PELTOR™ X3 Series Ear Muffs

Catalog No. p-7214252 Shop All 3M Products
Change view
Click to view available options
Model:
X3A
X3P3E
X3P5E
Dielectric:
No
Yes
Style:
Hard Hat
Over-the-head
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Dielectric Style
17-000-633 X3A Yes Over-the-head
17-000-638 X3P3E No Hard Hat
19-153-670 X3P5E Yes Hard Hat
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-000-633 Supplier 3M™ Supplier No. 7000104072

PELTOR™ X3 Series Ear Muffs in both over-the-head and hard hat-attached styles feature high attenuation ideal for moderate to high level noise environments.

  • Appropriate for moderate to high noise exposure
  • Twin headband design helps reduce heat buildup with good fit and balance
  • Ear cups ensure a customized fit and comfort for individual users
  • Convenient storage
  • Compatibility with hard hats (models X3P3E and X3P5E)
  • Dielectric (excludes model X3P3E)

Compliance

3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

TRUSTED_SUSTAINABILITY

Specifications

Color Black, Red
ANSI S3.1974 Yes
CSA Z94.2 Class AL
Cushion Color Black
For Use With (Application) Assembly, cleaning, demolition, facility maintenance, grinding, machine operations, painting, sanding, welding
Model X3A
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 28 dB
Product Type X3 Series
Headband Color Black
Dielectric Yes
Style Over-the-head
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.