Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pembrolizumab Mouse, Biotin, Clone: 10E12C6, GenScript™

Catalog No. 502120616
Encompass_Preferred
Change view
Click to view available options
Quantity:
40 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-212-0616 40 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-212-0616 Supplier Genscript Corporation Supplier No. A0184540
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute the lyophilized antibody with deionized water (or equivalent) to a final concentration of 0.5 mg/mL. Pembrolizumab (Keytruda) is a humanized monoclonal antibody that is approved by the U.S. Food and Drug Administration for the treatment of patients with unresectable or metastatic melanoma. Pembrolizumab binds to the human cell surface receptor PD-1 (Programmed Cell Death Protein 1) and blocks its interaction with PD-L1, resulting in the activation of T-cell-mediated immune responses against tumor cells. GenScript Anti-Pembrolizumab Antibody (10E12C6)[BIOTIN], mAb, Mouse is produced from a hybridoma resulting from the fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with Pembrolizumab.

Pembrolizumab Monoclonal antibody specifically detects Pembrolizumab in Chemical samples. It is validated for Blocking assay, ELISA

Specifications

Antigen Pembrolizumab
Applications Blocking Assay, ELISA
Classification Monoclonal
Clone 10E12C6
Conjugate Biotin
Formulation PBS with BSA and 0.02% sodium azide; pH 7.4
Gene Alias Keytruda
Host Species Mouse
Immunogen Pembrolizumab
Purification Method Protein A
Quantity 40 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Lyophilized
Isotype IgG1 κ
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.