Learn More
Pembrolizumab Rabbit, Biotin, Clone: 90G12, GenScript™
Rabbit Monoclonal Antibody
Supplier: Genscript Corporation A0185340
Description
Reconstitute the lyophilized antibody with deionized water (or equivalent) to a final concentration of 0.5 mg/mL. Pembrolizumab(Keytruda) is a humanized monoclonal antibody that is approved by the U.S. Foodand Drug Administration for the treatment of patients with unresectable ormetastatic melanoma. Pembrolizumab binds to the human cell surface receptorPD-1 (Programmed Cell Death Protein 1) and blocks its interaction with PD-L1,resulting in the activation of T-cell-mediated immune responses against tumorcells.GenScript MonoRabTM Anti-PembrolizumabAntibody (90G12F8) [Biotin], mAb, Rabbit is produced from ahybridoma resulting from the fusion of partner and B-lymphocytes obtained froma rabbit immunized with Pembrolizumab.
Pembrolizumab Monoclonal antibody specifically detects Pembrolizumab in Chemical samples. It is validated for Blocking assay, ELISASpecifications
| Pembrolizumab | |
| Monoclonal | |
| Biotin | |
| Keytruda | |
| Pembrolizumab | |
| 40 μg | |
| Primary | |
| Antibody | |
| IgG |
| Blocking Assay, ELISA | |
| 90G12 | |
| PBS with BSA and 0.02% sodium azide; pH 7.4 | |
| Rabbit | |
| Protein A | |
| RUO | |
| -20°C or -80°C if preferred | |
| Lyophilized | |
| Chemical |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.