Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pemetrexed disodium salt 2.5 hydrate, 98%, Thermo Scientific Chemicals

Catalog No. p-7077605
Click to view available options
Quantity:
1 g
5 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 357166-30-4
Molecular Formula C20H19N5Na2O6 . 5/2 H2O
Molecular Weight (g/mol) 516.43
MDL Number MFCD13194950
InChI Key NYDXNILOWQXUOF-CETBVFFHNA-L
Synonym pemetrexed disodium hemipenta hydrate
PubChem CID 131673885
IUPAC Name (2S)-2-[[4-[2-(2-amino-4-oxo-1,7-dihydropyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]amino]pentanedioic acid;molecular hydrogen;sodium;pentahydrate
SMILES [Na+].[Na+].NC1=NC2=C(C(CCC3=CC=C(C=C3)C(=O)N[C@@H](CCC([O-])=O)C([O-])=O)=CN2)C(=O)N1

Specifications

Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 516.43
Percent Purity 98%
Chemical Name or Material Pemetrexed disodium salt 2.5 hydrate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Suspected of causing genetic defects. May damage fertility. May damage the unborn child.
Precautionary Statement GHS P Statement Obtain special instructions before use. Use personal protective equipment as required. IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.