Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PEMT Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542383
Description
Peptide sequence: GWAIMHASPT GLLLTVLVAL TYIVALLYEE PFTAEIYRQK ASGSHKRS Sequence homology: Cow: 93%; Dog: 86%; Guinea Pig: 83%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 86%; Rabbit: 100%; Rat: 100%; Yeast: 100%; Zebrafish: 82%.
This gene encodes an enzyme which converts phosphatidylethanolamine to phosphatidylcholine by sequential methylation in the liver. The protein localizes to the endoplasmic reticulum and mitochondria-associated membranes. The gene is within the Smith-Magenis syndrome region on chromosome 17. Alternate splicing of this gene results in three transcript variants encoding two different isoforms.
Specifications
PEMT | |
Polyclonal | |
Unconjugated | |
PEMT | |
MGC2483; PEAMT; PEMPT; Pempt2; PEMT; PEMT2; PHOMETH; phosphatidylethanolamine N-methyltransferase; phosphatidylethanolamine N-methyltransferase-like protein; phospholipid methyltransferase; PLMT; PNMT; similar to phosphatidylethanolamine N-methyltransferase; zgc:55479 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10400, 25511 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot, Surface Plasmon Resonance | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q08388, Q9UBM1 | |
PEMT | |
Synthetic peptide directed towards the C-terminal of human PEMT (aa 152-199). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction