Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PEMT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559961
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559961 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559961 Supplier Invitrogen™ Supplier No. PA559961
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GFAGTFLGDY FGILKEARVT VFPFNILDNP MYWGSTANYL GWAIM Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.

This gene encodes an enzyme which converts phosphatidylethanolamine to phosphatidylcholine by sequential methylation in the liver. The protein localizes to the endoplasmic reticulum and mitochondria-associated membranes. The gene is within the Smith-Magenis syndrome region on chromosome 17. Alternate splicing of this gene results in three transcript variants encoding two different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEMT
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PEMT
Gene Accession No. Q9UBM1
Gene Alias MGC2483; PEAMT; PEMPT; Pempt2; PEMT; PEMT2; PHOMETH; phosphatidylethanolamine N-methyltransferase; phosphatidylethanolamine N-methyltransferase-like protein; phospholipid methyltransferase; PLMT; PNMT; similar to phosphatidylethanolamine N-methyltransferase; zgc:55479
Gene Symbols PEMT
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PEMT. Recombinant protein control fragment (Product #RP-91146).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10400
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.