Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PEMT Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5140153
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: LYWNVVARWE HKTRKLSRAF G.
This gene encodes an enzyme which converts phosphatidylethanolamine to phosphatidylcholine by sequential methylation in the liver. The protein localizes to the endoplasmic reticulum and mitochondria-associated membranes. The gene is within the Smith-Magenis syndrome region on chromosome 17. Alternate splicing of this gene results in three transcript variants encoding two different isoforms.
Specifications
| PEMT | |
| Polyclonal | |
| Unconjugated | |
| PEMT | |
| MGC2483; PEAMT; PEMPT; Pempt2; PEMT; PEMT2; PHOMETH; phosphatidylethanolamine N-methyltransferase; phosphatidylethanolamine N-methyltransferase-like protein; phospholipid methyltransferase; PLMT; PNMT; similar to phosphatidylethanolamine N-methyltransferase; zgc:55479 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10400 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UBM1 | |
| PEMT | |
| Recombinant protein corresponding to Human PEMT. Recombinant protein control fragment (Product #RP-109374). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction