Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PEN1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144453
Description
PEN1 Polyclonal Antibody for Western Blot, ELISA
Converts oxidosqualene to arabidiol. Minor production of arabidiol 20,21-epoxide.
Specifications
PEN1 | |
Polyclonal | |
Unconjugated | |
PEN1 | |
04C11 | |
Rabbit | |
Protein A/G | |
RUO | |
827200 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
2.5 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
Q9FR95 | |
PEN1 | |
Recombinant Arabidopsis thaliana PEN1 protein. | |
100 μL | |
Primary | |
Plant | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction