Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Penicillin Antibody (Pen-9), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314083AF532
This item is not returnable.
View return policy
Description
Penicillin Monoclonal antibody specifically detects Penicillin in All species samples. It is validated for ELISA, ImmunofluorescenceSpecifications
Penicillin | |
Monoclonal | |
Alexa Fluor 532 | |
Ampicillin, Benzylpenicillin, dicloxacillin, flucloxacillin, methicillin, PCN, Penicilin, Penicillin G, Phenoxymethylpenicillin | |
Penicilloyl-transferrin conjugate | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |
ELISA, Immunofluorescence | |
Pen-9 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
All species | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction