Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Penicillin Antibody (Pen-9), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314083F
This item is not returnable.
View return policy
Description
Penicillin Monoclonal antibody specifically detects Penicillin in All species samples. It is validated for ELISA, ImmunofluorescenceSpecifications
| Penicillin | |
| Monoclonal | |
| FITC | |
| Ampicillin, Benzylpenicillin, dicloxacillin, flucloxacillin, methicillin, PCN, Penicilin, Penicillin G, Phenoxymethylpenicillin | |
| Penicilloyl-transferrin conjugate | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| ELISA, Immunofluorescence | |
| Pen-9 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| All species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction