Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Penicillinase Polyclonal Antibody, Biotin
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA127255
Description
PA1-27255 detects Penicillinase from Enterobacter cloacae samples.

Specifications
Penicillinase | |
Polyclonal | |
Biotin | |
Beta lactamase; Oxacillinase | |
Full length protein from E. cloacae. | |
1 mg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Dot Blot, ELISA, Immunomicroscopy, Western Blot | |
10 mg/mL | |
PBS with 0.01% sodium azide; pH 7.2 | |
Rabbit | |
IgG fraction | |
RUO | |
Bacteria | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction