Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Microbiologics™ Penicillium venetum ATCC 16025

Click to view available options
Quantity:
2 Pellets
Description
- Simply rehydrate LYFO DISK qualitative microorganism pellets with a diluent, such as phosphate buffer or saline to inoculate culture media.
- Packaged in a glass vial containing 6 pellets of a single strain
Specifications
Specifications
| ATCC® | ATCC 16025 |
| Biosafety Level | 1 |
| Content And Storage | 2°C to 8°C |
| Format | Lyfo Disk |
| Manufacturing Origin | United States |
| Product Type | Quality Control |
| For Use With (Application) | Microbial Identification |
| Packaging Type | Plastic Canister |
| Quantity | 2 Pellets |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction