Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PENK Monoclonal Antibody (2181CT109.65.83)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA537792
Description
PENK Monoclonal Antibody for Western Blot
neuropeptide hormone; plays a role in the responses to stress and pain perception.
Specifications
PENK | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P01210 | |
PENK | |
recombinant protein from the human region of human PENK. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Western Blot | |
2181CT109.65.83 | |
Unconjugated | |
PENK | |
AI326464; ENK; enkephalin; enkephalin A; Leu-enkephalin; Met-enkephalin; Met-enkephalin-Arg-Gly-Leu; Met-enkephalin-Arg-Phe; Met-enkephalin-Arg-Ser-Leu; OGF; Opioid growth factor; PE; Penk; PENK(114-133); PENK(143-183); PENK(143-184); PENK(143-185); PENK(237-258); PENK(238-259); PENK(239-260); Penk1; Penk2; PENK-A; Penk-rs; PPA; preproenkephalin; preproenkephalin 1; preproenkephalin 2; proenkephalin; proenkephalin 1; proenkephalin 2; proenkephalin related sequence; proenkephalin-A; spermatogenic-specific proenkephalin; Synenkephalin | |
Mouse | |
Protein G | |
RUO | |
5179 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction