Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentachlorophenol Solution, 1000μg/mL in methanol, Ultra Scientific

Catalog No. usph1801
Encompass
Click to view available options
Quantity:
1 x 1 mL

Chemical Identifiers

CAS 87-86-5, 67-56-1
Molecular Formula C6HCl5O
Molecular Weight (g/mol) 266.323
InChI Key IZUPBVBPLAPZRR-UHFFFAOYSA-N
PubChem CID 992
ChEBI CHEBI:17642
IUPAC Name 2,3,4,5,6-pentachlorophenol
SMILES C1(=C(C(=C(C(=C1Cl)Cl)Cl)Cl)Cl)O

Specifications

CAS Max % 99.87, 0.13
Analyte or Component Names Methanol,pentachlorophenol
Element Methanol, Pentachlorophenol
Solvent or Matrix Methanol
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 1000μg/mL
Chemical Name or Material Pentachlorophenol Solution
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.