Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pentadecane, ≥99.8% (GC), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
5 mL
25 mL
Description
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications
Specifications
| CAS | 629-62-9 |
| Vapor Pressure | 1 mmHg (91.6°C) |
| Vapor Density | 7.4 (vs. air) |
| Density | 0.769 g/mL (at 25°C) |
| Flash Point | 132.00°C - Closed Cup |
| Physical Form | Neat |
| Linear Formula | CH3(CH2)13 CH3 |
| MDL Number | MFCD00008990 |
| Melting Point | 8°C to 10°C |
| Boiling Point | 270°C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction