Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentadecylcyclohexane 98.0+%, TCI America™
SDP

Catalog No. P036925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P036925G 25 g
P03695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P036925G Supplier TCI America Supplier No. P036925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6006-95-7
Molecular Formula C21H42
Molecular Weight (g/mol) 294.57
MDL Number MFCD00058952
InChI Key ZNUABQHWFGTOCO-UHFFFAOYSA-N
PubChem CID 22328
IUPAC Name pentadecylcyclohexane
SMILES CCCCCCCCCCCCCCCC1CCCCC1

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 294.57
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material Pentadecylcyclohexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.