Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentaerythritol, 98+%

Catalog No. AAA105200I
Click to view available options
Quantity:
250 g
1000 g
5000 g

CAS: 115-77-5 | C5H12O4 | 136.147 g/mol

Pentaerythritol is used as chemical intermediate for alkylated resins, synth lubricants, pentaerythritol tetranitrate & resin & tall oil esters. It is also used in coating, dynamite and plastic industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentaerythritol is used as chemical intermediate for alkylated resins, synth lubricants, pentaerythritol tetranitrate & resin & tall oil esters. It is also used in coating, dynamite and plastic industry.

Solubility
Soluble in Water: 0.1 g/mL

Notes
Hygroscopic. Incompatible with strong acids, strong oxidizing agents, acid chlorides, acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-77-5
Molecular Formula C5H12O4
Molecular Weight (g/mol) 136.147
MDL Number MFCD00004692
InChI Key WXZMFSXDPGVJKK-UHFFFAOYSA-N
Synonym pentaerythritol, pentek, 2,2-bis hydroxymethyl propane-1,3-diol, tetramethylolmethane, monopentaerythritol, auxinutril, maxinutril, monopentek, pentaerythrite, penetek
PubChem CID 8285
IUPAC Name 2,2-bis(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)CO)O

Specifications

Melting Point 260°C to 264°C
Density 1.396
Boiling Point 276°C (30 mmHg)
Flash Point 240°C (464°F)
Quantity 5000 g
Beilstein 1679274
Sensitivity Hygroscopic
Merck Index 14,7111
Solubility Information Soluble in Water: 0.1g/mL
Formula Weight 136.15
Percent Purity ≥98%
Chemical Name or Material Pentaerythritol
Show More Show Less
Hazard Category H500
EINECSNumber 204-104-9
RTECSNumber RZ2490000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.