Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentaerythritol, 98%, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
5kg
Packaging:
Plastic bottle
115-77-5
C5H12O4
136.15
WXZMFSXDPGVJKK-UHFFFAOYSA-N
pentaerythritol,pentek,2,2-bis hydroxymethyl propane-1,3-diol,tetramethylolmethane,monopentaerythritol,auxinutril,maxinutril,monopentek,pentaerythrite,penetek
8285
2,2-bis(hydroxymethyl)propane-1,3-diol
C(C(CO)(CO)CO)O
115-77-5
C5H12O4
136.15
WXZMFSXDPGVJKK-UHFFFAOYSA-N
pentaerythritol,pentek,2,2-bis hydroxymethyl propane-1,3-diol,tetramethylolmethane,monopentaerythritol,auxinutril,maxinutril,monopentek,pentaerythrite,penetek
8285
2,2-bis(hydroxymethyl)propane-1,3-diol
C(C(CO)(CO)CO)O
Melting Point 257.0°C to 264.0°C
Boiling Point 276.0°C (30.0 mmHg)
Quantity 250g
Flash Point 240°C
Infrared Spectrum Authentic
Color White
Packaging Plastic bottle
Assay Percent Range 98%
Linear Formula C(CH2OH)4
Beilstein 01,528
Merck Index 15,7221
Solubility Information Solubility in water: 1 g/18 mL water (15°C). Other solubilities: slightly soluble in alcohol and formamide, insoluble in benzene, carbon tetrachloride, ether,, acetone and petroleum ether
Formula Weight 136.15
Physical Form Crystals
Percent Purity 98%
Chemical Name or Material Pentaerythritol
Show More Show Less