Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentaerythritol, 98%

Catalog No. p-4509858
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic Drum
Plastic bottle

CAS: 115-77-5 | C5H12O4 | 136.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-77-5
Molecular Formula C5H12O4
Molecular Weight (g/mol) 136.15
InChI Key WXZMFSXDPGVJKK-UHFFFAOYSA-N
Synonym pentaerythritol, pentek, 2,2-bis hydroxymethyl propane-1,3-diol, tetramethylolmethane, monopentaerythritol, auxinutril, maxinutril, monopentek, pentaerythrite, penetek
PubChem CID 8285
IUPAC Name 2,2-bis(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)CO)O

Specifications

Melting Point 257.0°C to 264.0°C
Color White
Boiling Point 276.0°C (30.0 mmHg)
Flash Point 240°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Linear Formula C(CH2OH)4
Quantity 250 g
Beilstein 01,528
Merck Index 15,7221
Solubility Information Solubility in water: 1 g/18 mL water (15°C). Other solubilities: slightly soluble in alcohol and formamide, insoluble in benzene, carbon tetrachloride, ether,, acetone and petroleum ether
Formula Weight 136.15
Percent Purity 98%
Physical Form Crystals
Chemical Name or Material Pentaerythritol
Show More Show Less
EINECSNumber 204-104-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.