Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentaethylene glycol, 98+%

Catalog No. p-4408455
Click to view available options
Quantity:
10 g
50 g
250 g

Used as an intermediate in the synthesis of cyclic and noncyclic polyethers, complexing agents and immobilized phase-transfer catalysts. It acts as a catalyst for potassium hydroxide induced elimination reactions in benzene. Further, it is used in functional fluids and viscosity adjustors. | CAS: 4792-15-8 | C10H22O6 | 238.28 g/mol

Pentaethylene glycol is used as an intermediate in the synthesis of cyclic and noncyclic polyethers, complexing agents and immobilized phase-transfer catalysts. It acts as a catalyst for potassium hydroxide induced elimination reactions in benzene. Further, it is used in functional fluids and viscosity adjustors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentaethylene glycol is used as an intermediate in the synthesis of cyclic and noncyclic polyethers, complexing agents and immobilized phase-transfer catalysts. It acts as a catalyst for potassium hydroxide induced elimination reactions in benzene. Further, it is used in functional fluids and viscosity adjustors.

Solubility
Miscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, acid chlorides and acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4792-15-8
Molecular Formula C10H22O6
Molecular Weight (g/mol) 238.28
MDL Number MFCD00002878
InChI Key JLFNLZLINWHATN-UHFFFAOYSA-N
Synonym pentaethylene glycol, 3,6,9,12-tetraoxatetradecane-1,14-diol, pentaethyleneglycol, pentaglycol, ho ch2ch2o 5h, 3,6,9,12-tetraoxatetradocane-1,14-diol, 2-2-2-2-2-hydroxyethoxy ethoxy ethoxy ethoxy ethanol, dsstox_cid_7579, dsstox_rid_78510, dsstox_gsid_27579
PubChem CID 62551
ChEBI CHEBI:39631
IUPAC Name 2-[2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethoxy]ethanol
SMILES C(COCCOCCOCCOCCO)O

Specifications

Density 1.126 g/mL
Boiling Point 338°C to 340°C
Flash Point >110°C (230°F)
Assay Percent Range >98%
Refractive Index 1.462
Quantity 10 g
Beilstein 1635593
Solubility Information Miscible with water.
Formula Weight 238.28
Chemical Name or Material Pentaethylene glycol
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 225-341-4
RTECSNumber RZ2670000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.