Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluorobenzamide 99.0+%, TCI America™
SDP

Catalog No. P092925G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 652-31-3
Molecular Formula C7H2F5NO
Molecular Weight (g/mol) 211.091
MDL Number MFCD00007971
InChI Key WPWWHXPRJFDTTJ-UHFFFAOYSA-N
PubChem CID 69547
IUPAC Name 2,3,4,5,6-pentafluorobenzamide
SMILES C1(=C(C(=C(C(=C1F)F)F)F)F)C(=O)N

Specifications

Melting Point 148°C
Color White
Quantity 25 g
Synonym pentafluorobenzamide, benzamide, 2,3,4,5,6-pentafluoro, benzamide, pentafluoro, 2,3,4,5,6-pentafluoro-benzamide, acmc-209nq0, benzamide,3,4,5,6-pentafluoro, benzamide,2,3,4,5,6-pentafluoro
Formula Weight 211.09
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Pentafluorobenzamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.