Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluorobenzene, 98+%

Catalog No. AAA1394814
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1394814 25 g
AAA1394806 5 g
AAA1394822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1394814 Supplier Thermo Scientific Chemicals Supplier No. A1394814
Add to Cart

CAS: 363-72-4 | C6HF5 | 168.07 g/mol

Pentafluorobenzene is a fluorinated benzene with anesthetic properties. Pentafluorobenzene has been shown to potently and reversibly inhibited human α4β2 neuronal nicotinic acetylcholine receptor depe ndent on drug hydrophobicity in Xenopus laevis oocyte. New reaction conditions are described that enable the direct arylation of pentafluorobenzene with sterically encumbered aryl bromides and aryl chlorides. Bis (pentafluoropheny1) methane and tris (pentafluoropheny1) methane were prepared in 77 and 92% yields, respectively, by a Friedel-Crafts reaction of pentafluorobenzene with methylene chloride and chloroform using aluminum chloride as the Lewis acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentafluorobenzene is a fluorinated benzene with anesthetic properties. Pentafluorobenzene has been shown to potently and reversibly inhibited human α4β2 neuronal nicotinic acetylcholine receptor depe ndent on drug hydrophobicity in Xenopus laevis oocyte. New reaction conditions are described that enable the direct arylation of pentafluorobenzene with sterically encumbered aryl bromides and aryl chlorides. Bis (pentafluoropheny1) methane and tris (pentafluoropheny1) methane were prepared in 77 and 92% yields, respectively, by a Friedel-Crafts reaction of pentafluorobenzene with methylene chloride and chloroform using aluminum chloride as the Lewis acid.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 363-72-4
Molecular Formula C6HF5
Molecular Weight (g/mol) 168.07
MDL Number MFCD00000286
InChI Key WACNXHCZHTVBJM-UHFFFAOYSA-N
Synonym pentafluorobenzene, benzene, pentafluoro, unii-ih36lu53xs, benzene, 1,2,3,4,5-pentafluoro, 2,3,4,5,6-pentafluorobenzene, ih36lu53xs, 1,2,3,4,5-pentafluoro-benzene, pentafluorophenyl, benzenepentafluoride, pubchem2310
PubChem CID 9696
IUPAC Name 1,2,3,4,5-pentafluorobenzene
SMILES FC1=CC(F)=C(F)C(F)=C1F

Specifications

Melting Point -48°C
Density 1.518
Boiling Point 84°C to 85°C
Flash Point 13°C (55°F)
Refractive Index 1.391
Quantity 25 g
UN Number UN1993
Beilstein 1911549
Solubility Information Insoluble in water.
Formula Weight 168.06
Percent Purity ≥98%
Chemical Name or Material Pentafluorobenzene
Show More Show Less
Hazard Category H225-H302-H315-H319-H335
Hazard Statement GHS H Statement
H225-H302
Highly flammable liquid and vapour.
Harmful if swallowed.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 206-658-7
RTECSNumber DA6651600
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.