Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluorobenzenesulfonyl chloride, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
832-53-1
C6ClF5O2S
266.57
MFCD00007427
UOJCTEGNHXRPKO-UHFFFAOYSA-N
pentafluorobenzenesulfonyl chloride,2,3,4,5,6-pentafluorobenzene-1-sulfonyl chloride,pentafluorobenzenesulphonyl chloride,benzenesulfonyl chloride, pentafluoro,pentafluorophenylsulfonyl chloride,2,3,4,5,6-pentafluorobenzenesulfonyl choride,2,3,4,5,6-pentafluoro-benzenesulfonyl chloride,chloro 2,3,4,5,6-pentafluorophenyl sulfone,pubchem5683,pentafluorobenzenesulfonylchloride
70026
2,3,4,5,6-pentafluorobenzenesulfonyl chloride
C1(=C(C(=C(C(=C1F)F)S(=O)(=O)Cl)F)F)F
832-53-1
C6ClF5O2S
266.57
MFCD00007427
UOJCTEGNHXRPKO-UHFFFAOYSA-N
pentafluorobenzenesulfonyl chloride,2,3,4,5,6-pentafluorobenzene-1-sulfonyl chloride,pentafluorobenzenesulphonyl chloride,benzenesulfonyl chloride, pentafluoro,pentafluorophenylsulfonyl chloride,2,3,4,5,6-pentafluorobenzenesulfonyl choride,2,3,4,5,6-pentafluoro-benzenesulfonyl chloride,chloro 2,3,4,5,6-pentafluorophenyl sulfone,pubchem5683,pentafluorobenzenesulfonylchloride
70026
2,3,4,5,6-pentafluorobenzenesulfonyl chloride
C1(=C(C(=C(C(=C1F)F)S(=O)(=O)Cl)F)F)F
Quantity 25g
Boiling Point 211°C
Flash Point 90°C
Specific Gravity 1.796
Refractive Index 1.4780 to 1.48
Packaging Glass bottle
Density 1.7960g/mL
Assay Percent Range 98.5% min. (GC)
Linear Formula C6F5SO2Cl
Formula Weight 266.57
Percent Purity 99%
Chemical Name or Material Pentafluorobenzenesulfonyl chloride, 99%
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 212-620-0