Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluorobenzoyl Chloride 98.0+%, TCI America™
SDP

Catalog No. P080725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P080725G 25 g
P08075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P080725G Supplier TCI America Supplier No. P080725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2251-50-5
Molecular Formula C7ClF5O
Molecular Weight (g/mol) 230.52
MDL Number MFCD00000657
InChI Key MYHOHFDYWMPGJY-UHFFFAOYSA-N
Synonym pentafluorobenzoyl chloride, perfluorobenzoyl chloride, benzoyl chloride, pentafluoro, pentafluorobenzoylchloride, pubchem15522, acmc-1covi, intermediates-zcf02601, pentafluorobenzoic acid chloride, attercop-chm at133861, 67924-15-6 unspecified acetate
PubChem CID 75256
ChEBI CHEBI:39425
IUPAC Name pentafluorobenzoyl chloride
SMILES FC1=C(F)C(F)=C(C(Cl)=O)C(F)=C1F

Specifications

Color Yellow
Boiling Point 159°C
Quantity 25 g
UN Number 3265
Formula Weight 230.52
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Pentafluorobenzoyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.