Learn More
Pentafluoroiodobenzene, 97%
Description
Iodopentafluorobenzene was used to study the formation of radical ions of iodopentafluorobenzene in aqueous solution. It was used as solvent in a study to determine singlet oxygen lifetimes from phosphorescence decays in halogen substituted perfluorinated solvents by infrared emission spectrometery. It has potential applications in plasma processing industry and in preparation of catalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Iodopentafluorobenzene was used to study the formation of radical ions of iodopentafluorobenzene in aqueous solution. It was used as solvent in a study to determine singlet oxygen lifetimes from phosphorescence decays in halogen substituted perfluorinated solvents by infrared emission spectrometery. It has potential applications in plasma processing industry and in preparation of catalysts.
Solubility
Insoluble in water.
Notes
Light sensitive. Store at room temperature. Keep away from oxidizing agents.
Specifications
Specifications
| Melting Point | -29°C |
| Density | 2.212 |
| Boiling Point | 161°C to 163°C |
| Refractive Index | 1.496 |
| Quantity | 50 g |
| Beilstein | 2051549 |
| Sensitivity | Light sensitive |
| Solubility Information | Insoluble in water. |
| Formula Weight | 293.96 |
| Percent Purity | 97% |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 212-565-2 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.