Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Toluoyl chloride, 98%

Catalog No. AC147980010
Click to view available options
Quantity:
5 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle

CAS: 874-60-2 | C8H7ClO | 154.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874-60-2
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.6
MDL Number MFCD00000696
InChI Key NQUVCRCCRXRJCK-UHFFFAOYSA-N
Synonym p-toluoyl chloride, benzoyl chloride, 4-methyl, p-methylbenzoyl chloride, 4-toluoyl chloride, p-toluyl chloride, p-toluic acid chloride, 4-methylbenzoic acid chloride, 4-methylbenzoylchloride, 4-methyl-benzoyl chloride, unii-092e36pwl4
PubChem CID 13405
IUPAC Name 4-methylbenzoyl chloride
SMILES CC1=CC=C(C=C1)C(=O)Cl

Specifications

Melting Point -2.0°C
Color Colorless to Brown
Density 1.1600g/mL
Boiling Point 225.0°C to 227.0°C
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.5525 to 1.5545
Linear Formula CH3C6H4COCl
Quantity 1 L
Beilstein 09,486
Specific Gravity 1.16
Solubility Information Solubility in water: decomposes.
Formula Weight 154.6
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material p-Toluoyl chloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 212-864-8
RTECSNumber XV1660000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.