Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluorophenylhydrazine 98.0+%, TCI America™
SDP

Catalog No. P085225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P085225G 25 g
P08525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P085225G Supplier TCI America Supplier No. P085225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 828-73-9
Molecular Formula C6H3F5N2
Molecular Weight (g/mol) 198.096
MDL Number MFCD00007574
InChI Key BYCUWCJUPSUFBX-UHFFFAOYSA-N
Synonym pentafluorophenylhydrazine, pentafluorophenyl hydrazine, perfluorophenyl hydrazine, hydrazine, pentafluorophenyl, 2,3,4,5,6-pentafluorophenyl hydrazine, 1-2,3,4,5,6-pentafluorophenyl hydrazine, pubchem3316, pentafluorophenyl-hydrazine
PubChem CID 13236
IUPAC Name (2,3,4,5,6-pentafluorophenyl)hydrazine
SMILES C1(=C(C(=C(C(=C1F)F)F)F)F)NN

Specifications

Melting Point 76°C
Quantity 25 g
Formula Weight 198.10
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pentafluorophenylhydrazine
RTECSNumber MV8325000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.