Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluoropropionic Acid 98.0+%, TCI America™
SDP

Catalog No. P1125100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1125100G 100 g
P112525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1125100G Supplier TCI America Supplier No. P1125100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 422-64-0
Molecular Formula C3HF5O2
Molecular Weight (g/mol) 164.03
MDL Number MFCD00004170
InChI Key LRMSQVBRUNSOJL-UHFFFAOYSA-N
Synonym pentafluoropropionic acid, perfluoropropionic acid, pentafluoropropanoic acid, propanoic acid, pentafluoro, propionic acid, pentafluoro, 2,2,3,3,3-pentafluoropropionic acid, ipc-pffa-3, perfluoropropanoic acid, propanoic acid, 2,2,3,3,3-pentafluoro, perfluoropropionicacid
PubChem CID 62356
IUPAC Name pentafluoropropanoic acid
SMILES OC(=O)C(F)(F)C(F)(F)F

Specifications

Color Yellow
Boiling Point 98°C
Quantity 100 g
UN Number 3265
Formula Weight 164.03
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Pentafluoropropionic Acid
EINECSNumber (2)-1182
RTECSNumber UF6475000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.