Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluorothiophenol, 97%

Catalog No. AAB2232314
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 771-62-0 | C6HF5S | 200.13 g/mol

Pentafluorothiophenol is used as a fluorinated building block

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentafluorothiophenol is used as a fluorinated building block

Solubility
Insoluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 771-62-0
Molecular Formula C6HF5S
Molecular Weight (g/mol) 200.13
MDL Number MFCD00004828
InChI Key UVAMFBJPMUMURT-UHFFFAOYSA-N
Synonym pentafluorothiophenol, pentafluorobenzenethiol, benzenethiol, pentafluoro, pentafluorophenyl mercaptan, 2,3,4,5,6-pentafluorothiophenol, 2,3,4,5,6-pentafluorobenzene-1-thiol, pentafluorobenzene thiol, pentafluorobenzene-1-thiol, 2,3,4,5,6-pentafluorothiphenol
PubChem CID 13042
IUPAC Name 2,3,4,5,6-pentafluorobenzenethiol
SMILES FC1=C(F)C(F)=C(S)C(F)=C1F

Specifications

Melting Point -24°C
Density 1.572
Boiling Point 143°C
Flash Point 51°C (123°F)
Odor Stench
Refractive Index 1.465
Quantity 25 g
UN Number UN2920
Beilstein 1876292
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 200.13
Percent Purity 97%
Chemical Name or Material Pentafluorothiophenol
Show More Show Less
missing translation for 'healthHazard1' H226-H302+H312-H314-H331-H335
missing translation for 'healthHazard2' GHS H Statement
H314-H318-H226-H302-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful if inhaled.
missing translation for 'healthHazard3' P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber 212-236-3
RTECSNumber DC1940000
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.