Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentamethylbenzene 99.0+%, TCI America™
SDP

Catalog No. P004325G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 700-12-9
Molecular Formula C11H16
Molecular Weight (g/mol) 148.249
MDL Number MFCD00008522
InChI Key BEZDDPMMPIDMGJ-UHFFFAOYSA-N
Synonym pentamethylbenzene, benzene, pentamethyl, pentamethylbenzol, benzene, 1,2,3,4,5-pentamethyl, pentamethyl phenyl, acmc-1bkt8, ksc492e3d, 1,3,4,5-pentamethylbenzene, pentamethylbenzene 5g
PubChem CID 12784
ChEBI CHEBI:38998
IUPAC Name 1,2,3,4,5-pentamethylbenzene
SMILES CC1=CC(=C(C(=C1C)C)C)C

Specifications

Melting Point 53°C
Color White
Boiling Point 231°C
Quantity 25 g
UN Number 1325
Formula Weight 148.25
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Pentamethylbenzene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.