Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pentamethylcyclopentadienylmolybdenum dicarbonyl dimer, 99%
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 219°C (decomposition) |
| Quantity | 5 g |
| UN Number | UN3466 |
| Formula Weight | 574.38 |
| Percent Purity | 99% |
| Physical Form | Crystalline |
| Chemical Name or Material | Pentamethylcyclopentadienylmolybdenum dicarbonyl dimer |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement H300-H330 Fatal if swallowed. Fatal if inhaled. |
| Precautionary Statement | GHS P Statement P260-P301+P310-P304+P340-P320-P330-P405-P501a Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Specific treatment is urgent (see label). Rinse mouth. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S. |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction