Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentamethylcyclopentadienylmolybdenum dicarbonyl dimer, 99%

Catalog No. p-7050703
Click to view available options
Quantity:
250 mg
1 g
5 g

CAS: 12132-04-6 | C12H16MoO2 | 288.208 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12132-04-6
Molecular Formula C12H16MoO2
Molecular Weight (g/mol) 288.208
MDL Number MFCD00058846
InChI Key CIKWUWWMMYQFKN-UHFFFAOYSA-N
Synonym Dicarbonyl(pentamethylcyclopentadienyl)molybdenum dimer
PubChem CID 131675733
IUPAC Name carbon monoxide;molybdenum;1,2,3,4,5-pentamethylcyclopenta-1,3-diene
SMILES CC1C(=C(C(=C1C)C)C)C.[C-]#[O+].[C-]#[O+].[Mo]

Specifications

Melting Point 219°C (decomposition)
Quantity 250 mg
UN Number UN3466
Formula Weight 574.38
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material Pentamethylcyclopentadienylmolybdenum dicarbonyl dimer
Hazard Category H300+H330
Hazard Statement GHS H Statement
H300-H330
Fatal if swallowed.
Fatal if inhaled.
Precautionary Statement P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.