Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentane CHROMASOLV™, for HPLC, ≥99.0%, Solstice

Catalog No. 6001114 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
4 x 4 L
6 x 1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-011-14 1 L
60-011-17 6 x 1 L
60-011-16 4 x 2.5 L
60-048-243 4 x 4 L
60-011-15 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Repeated exposure may cause skin dryness or cracking.
Catalog No. 60-011-14 Supplier Solstice Supplier No. 349561L
Add to Cart
Add to Cart
Repeated exposure may cause skin dryness or cracking.

CHROMASOLV™, for HPLC,≥99.0%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC

Specifications

Melting Point -130°C
Density 0.626g/mL at 25°C
Quantity 1 L
Refractive Index n20/D 1.358
Linear Formula CH3(CH2)3CH3
Beilstein 969132
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
Boiling Point 35°C to 36°C
Formula Weight 72.15g/mol
Percent Purity ≥99%
Grade CHROMASOLV
Chemical Name or Material Pentane, for HPLC
Show More Show Less
Hazard Statement P210-P261-P273-P301 + P310-P331
Precautionary Statement H225-H304-H336-H411
EINECSNumber 203-692-4
RTECSNumber RZ9450000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.