Learn More
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Specifications
Specifications
| Melting Point | -130°C |
| Density | 0.626g/mL at 25°C |
| Quantity | 4 x 2.5 L |
| Refractive Index | n20/D 1.358 |
| Linear Formula | CH3(CH2)3CH3 |
| Beilstein | 969132 |
| Synonym | n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish |
| Boiling Point | 35°C to 36°C |
| Formula Weight | 72.15g/mol |
| Percent Purity | ≥99% |
| Show More |
Safety and Handling
| Hazard Statement | P210-P261-P273-P301 + P310-P331 |
| Precautionary Statement | H225-H304-H336-H411 |
| EINECSNumber | 203-692-4 |
| RTECSNumber | RZ9450000 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.