Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pentane, CHROMASOLV™, for pesticide residue analysis, Honeywell Riedel-de Haën™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
Description
- For gas chromatography (GC)
- Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
- Non-volatile matter and water content are specified for each solvent

Specifications
Specifications
Density | 0.626g/mL at 25°C |
Quantity | 1 L |
Explosion Limit | 0.083 |
Flash Point | −49°C (−56.2°F) |
Ignition Point | 500°F |
Linear Formula | CH3(CH2)3CH3 |
UN Number | UN1265 |
Beilstein | 969132 |
Formula Weight | 72.15g/mol |
Percent Purity | ≥99% (GC) |
Show More |
Safety and Handling
Hazard Statement | P210-P261-P273-P301 + P310-P331 |
RTECSNumber | RZ9450000 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction