Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentoxifylline, BP, 99-101%, Spectrum™ Chemical
SDP

Catalog No. 18605003 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
2.5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail

P1196, 6493-05-6, C13H18N4O3

Spectrum™ Chemical Pentoxifylline, BP is a xanthine derivative used to treat the symptoms of peripheral artery disease. The BP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical BP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 6493-05-6
Molecular Formula C13H18N4O3
Molecular Weight (g/mol) 278.31
InChI Key BYPFEZZEUUWMEJ-UHFFFAOYSA-N
IUPAC Name 3,7-dimethyl-1-(5-oxohexyl)-2,3,6,7-tetrahydro-1H-purine-2,6-dione
SMILES CN1C=NC2=C1C(=O)N(CCCCC(C)=O)C(=O)N2C

Specifications

Acidity To pass test
CAS Max % 1
Loss on Drying 0.005
Packaging Amber Glass Bottle
Quantity 5 g
Percent Purity 99 to 101%
Grade BP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.