Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pentoxifylline, BP, 99-101%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
5 g
25 g
100 g
500 g
2.5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
Description
Spectrum™ Chemical Pentoxifylline, BP is a xanthine derivative used to treat the symptoms of peripheral artery disease. The BP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical BP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
Acidity | To pass test |
CAS Max % | 1 |
Loss on Drying | 0.005 |
Packaging | Poly Pail |
Quantity | 2.5 kg |
Percent Purity | 99 to 101% |
Grade | BP |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction