Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pentraxin 2/SAP Antibody (APCS/3240), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279848AF532
Description
Pentraxin 2/SAP Monoclonal specifically detects Pentraxin 2/SAP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Pentraxin 2/SAP | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
APCS | |
Recombinant fragment of human Pentraxin 2/SAP protein (around aa 143-223) (exact sequence is proprietary) (Uniprot: P02743) | |
0.1 mL | |
Neuroscience | |
Human | |
IgG2b κ |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
APCS/3240 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
9.5S alpha-1-glycoprotein, amyloid P component, serum, MGC88159, PTX2serum amyloid P-component, SAPpentaxin-related, Serum Amyloid P component | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction