Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentraxin 2/SAP Antibody (OTI1D6) - Azide and BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP273334

Catalog No. NBP273334


Only null left
Add to Cart

Description

Description

Pentraxin 2/SAP Monoclonal specifically detects Pentraxin 2/SAP in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Pentraxin 2/SAP
Monoclonal
LYOPH
Western Blot 1:500-2000, Immunocytochemistry/Immunofluorescence 1:100
9.5S alpha-1-glycoprotein, amyloid P component, serum, MGC88159, PTX2serum amyloid P-component, SAPpentaxin-related, Serum Amyloid P component
Mouse
23.2 kDa
100 μg
Primary
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Store at -20°C. Avoid freeze-thaw cycles.
IgG2a
Western Blot, Immunocytochemistry, Immunofluorescence
OTI1D6
Unconjugated
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
APCS
Full length human recombinant protein of human APCS(NP_001630) produced in HEK293T cell.
Affinity Purified
Neuroscience
325
Human
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.