Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pentraxin 2/SAP, Mouse anti-Human, Clone: APCS/3240, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP27971420UG
Description
Pentraxin 2/SAP Monoclonal specifically detects Pentraxin 2/SAP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
Pentraxin 2/SAP | |
Monoclonal | |
0.2 mg/mL | |
Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
P02743, P02743 | |
APCS | |
Recombinant fragment of human Pentraxin 2/SAP protein (around aa 143-223) (exact sequence is proprietary) | |
20 μg | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
APCS/3240 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
9.5S alpha-1-glycoprotein, amyloid P component, serum, MGC88159, PTX2serum amyloid P-component, SAPpentaxin-related, Serum Amyloid P component | |
Mouse | |
Protein A or G purified | |
RUO | |
325 | |
Store at 4°C. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction