Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pentraxin 3/TSG-14 Antibody (10D9), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Pentraxin 3/TSG-14 |
---|---|
Clone | 10D9 |
Dilution | Flow Cytometry 1:50-1:200, ELISA |
Applications | Flow Cytometry, ELISA |
Classification | Monoclonal |
Description
Pentraxin 3/TSG-14 Monoclonal antibody specifically detects Pentraxin 3/TSG-14 in Human samples. It is validated for Flow Cytometry, ELISASpecifications
Pentraxin 3/TSG-14 | |
Flow Cytometry 1:50-1:200, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
alpha-induced protein 5, pentaxin-related gene, rapidly induced by IL-1 beta, tumor necrosis factor, Pentaxin-related protein PTX3, pentraxin 3, long, pentraxin-3, pentraxin-related gene, rapidly induced by IL-1 beta, pentraxin-related protein PTX3, TNF alpha-induced protein 5, TNFAIP5, TSG14, TSG-14pentaxin-related gene, rapidly induced by IL-1 beta, Tumor necrosis factor alpha-induced protein 5, tumor necrosis factor, alpha-induced protein 5, Tumor necrosis factor-inducible gene 14 protein, tumor necrosis factor-inducible protein TSG-14 | |
A synthesized peptide derived from Human Pentraxin 3/TSG-14 [UniProt P26022] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
10D9 | |
Flow Cytometry, ELISA | |
Unconjugated | |
Rabbit | |
Immunology | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
5806 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title