Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Pentylenetetrazole
GREENER_CHOICESDP

Catalog No. 26871G Shop All Novus Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-871-G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 26-871-G Supplier R&D Systems™ Supplier No. 2687/1G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. CNS stimulant

CNS stimulant that induces kindling in vivo. Causes alterations in excitatory and inhibitory neurotransmitter systems.

Chemical Identifiers

CAS 54-95-5
Molecular Formula C6H10N4

Specifications

Quantity 1 g
Product Line Tocris
Formula Weight 138.17
Purity >99%
Molecular Weight (g/mol) 138.174
Chemical Name or Material Pentylenetetrazole
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.