Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PEP12 Mouse anti-Yeast, Clone: 2C3G4, Abnova™

Catalog No. 89035536 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-536 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-536 Supplier Abnova Corporation Supplier No. MAB14138

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against yeast PEP12.

  • High specificity
  • High reactivity

Specifications

Antigen PEP12
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone 2C3G4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against yeast PEP12.
Dilution Immunofluorescence Western Blot (0.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In HEPES (0.02% sodium azide).
Gene Accession No. P32854
Gene Alias VPL6/VPS6/VPT13
Gene Symbols PEP12
Host Species Mouse
Immunogen Yeast Pep12p.
Purification Method Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 854201
Target Species Yeast
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.