Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PEPA, Tocris Bioscience™

Catalog No. 292950
Click to view available options
:
50mg
This item is not returnable. View return policy

Catalog No. 29-295-0

Supplier: R&D Systems 2929/50

This item is not returnable. View return policy

Allosteric potentiator of AMPA receptor desensitization

Novel allosteric potentiator of AMPA receptor desensitization. Slows the rate of onset of desensitization and potentiates steady-state equilibrium currents induced by glutamate with subunit (GluA3/4 > GluA1) and splice variant (flop > flip) selectivity. Ameliorates post-ischemic memory impairment in rats following i.v. administration.

Specifications

CAS 141286-78-4
Molecular Formula C16H16F2N2O4S2
Quantity 50 mg
Synonym pepa, 2,6-difluoro-4-2-phenylsulfonylamino ethylthio phenoxyacetamide, 2-2,6-difluoro-4-2-phenylsulfonyl amino ethyl sulfanyl phenoxy acetamide, lopac-d-8941, pepa drug, 4-2-phenylsulfonylamino ethylthio-2,6-difluorophenoxyacetamide, 4-2-phenylsulfonylamino ethylthio-2,6-difluoropheno-xyacetamide, 2,6-difluoro-4-2-phenylsul-fonyl-amino-ethyl-thio-phenoxy-acet-amide, 2,6-difluoro-4-2-phenylsul-fonyl-amino-ethyl-thio-phenoxy-acetamide, 2-2,6-difluoro-4-2-phenylsulfonyl amino ethyl thio phenoxy acetamide
InChI Key GTACSIONMHMRPD-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)S(=O)(=O)NCCSC2=CC(=C(C(=C2)F)OCC(=O)N)F
IUPAC Name 2-[4-[2-(benzenesulfonamido)ethylsulfanyl]-2,6-difluorophenoxy]acetamide
Molecular Weight (g/mol) 402.431
PubChem CID 6603828
Formula Weight 402.44
Percent Purity >99%
Chemical Name or Material PEPA
Show More Show Less
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.