Learn More
Invitrogen™ PCK2 Polyclonal Antibody

Description
This antibody is predicted to react with bovine, canine and porcine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.
Specifications
Specifications
| Antigen | PCK2 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | PCK2 |
| Gene Accession No. | Q16822, Q8BH04 |
| Gene Alias | 1810010O14Rik; 9130022B02Rik; HGNC:8725; PCK2; PEP carboxykinase; PEPCK; PEPCK2; PEPCK-M; Phosphoenolpyruvate carboxykinase [GTP], mitochondrial; phosphoenolpyruvate carboxykinase 2 (mitochondrial); phosphoenolpyruvate carboxykinase 2, mitochondrial; phosphoenolpyruvate carboxylase; phosphopyruvate carboxylase |
| Gene Symbols | PCK2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.