Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PEPCK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597059
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597059 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597059 Supplier Invitrogen™ Supplier No. PA597059
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAALYRPGLR LNWHGLSPLG WPSCRSIQTL RVLSGDLGQL PTGIRDFVEH SARLCQPEGI HICDGTEAEN TATLTLLEQQ GLIRKLPKYN NCWLARTDPK DVARVESKTV IVTPSQRDTV QLPPGGARGQ LGNWMSPADF QRAVDERFPG CMQGRTMYVL PFSMGPVGSP LSRIGVQLTD SAYVVASMRI MTRLGTPVLQ ALGDGDFVKC LHSVGQPLTG QGEPVSQWPC NPEKTLIGHV PDQREIISFG SGYGGNSLLG KKCFALRIAS RLARDEGWLA; Positive Samples: SH-SY5Y, LO2, HepG2, Mouse testis, Mouse brain, Mouse kidney, Rat brain; Cellular Location: Mitochondrion.

This gene encodes a member of the phosphoenolpyruvate carboxykinase (GTP) family. The protein is a mitochondrial enzyme that catalyzes the conversion of oxaloacetate to phosphoenolpyruvate in the presence of GTP. A cytosolic form encoded by a different gene has also been characterized and is the key enzyme of gluconeogenesis in the liver. The encoded protein may serve a similar function, although it is constitutively expressed and not modulated by hormones such as glucagon and insulin that regulate the cytosolic form. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEPCK2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.38 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PCK2
Gene Accession No. Q16822, Q8BH04
Gene Alias 1810010O14Rik; 9130022B02Rik; HGNC:8725; PCK2; PEP carboxykinase; PEPCK; PEPCK2; PEPCK-M; Phosphoenolpyruvate carboxykinase [GTP], mitochondrial; phosphoenolpyruvate carboxykinase 2 (mitochondrial); phosphoenolpyruvate carboxykinase 2, mitochondrial; phosphoenolpyruvate carboxylase; phosphopyruvate carboxylase
Gene Symbols PCK2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human PEPCK/PEPCK/PCK2 (NP_0045542).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361042, 5106, 74551
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.