Learn More
PEPD Mouse anti-Human, Clone: OTI1B7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501727

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Xaa-Pro dipeptidase is a cytosolic dipeptidase that hydrolyzes dipeptides with proline or hydroxyproline at the carboxy terminus (but not Pro-Pro). It is important in collagen metabolism because of the high levels of iminoacids.Xaa-Pro dipeptidase is a cytosolic dipeptidase that hydrolyzes dipeptides with proline or hydroxyproline at the carboxy terminus (but not Pro-Pro). It is important in collagen metabolism because of the high levels of iminoacids.Specifications
PEPD | |
Monoclonal | |
Unconjugated | |
PEPD | |
PROLIDASE | |
Mouse | |
Affinity Chromatography | |
RUO | |
5184 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI1B7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P12955 | |
PEPD | |
Full length human recombit protein of human PEPD produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.