Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PEPP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557463
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557463 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557463 Supplier Invitrogen™ Supplier No. PA557463
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PNGVNSVEMM DKERNKDKMP EDVTFSPQDE TQTANHKPEE HPEENTKNSV DEQEETVISY ESTPEVSRGN QTMAVKSLSP SPESSASPVP STQP Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 48%.

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H3 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEPP2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PLEKHA5
Gene Accession No. Q9HAU0
Gene Alias 2810431N21Rik; AI428202; AK129423; AMH-Cre; Ayu21-9; Gt(Ayu21)9Imeg; Gt(pU21)9Imeg; KIAA1686; PEPP2; PEPP-2; PH domain-containing family A member 5; Phosphoinositol 3-phosphate-binding protein 2; phosphoinositol 3-phosphate-binding protein-2; pleckstrin homology domain containing A5; pleckstrin homology domain containing, family A member 5; pleckstrin homology domain-containing family A member 5; PLEKHA5; Tg(AMH-cre)1Flor; TRS1
Gene Symbols PLEKHA5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PEPP2. Recombinant protein control fragment (Product #RP-97625).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54477
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.