Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Pepsin A (Porcine), 2X

Catalog No. 89956426
Click to view available options
Quantity:
1 g

Used for Enzyme

#REF!

Specifications

Accession Number P00791
For Use With (Application) Enzyme Activity
Gene ID (Entrez) 396892
Molecular Weight (g/mol) 34.6kDa
Name Pepsin A (Porcine), 2x
Preparation Method Native protein purified from Porcine Stomach
Quantity 1 g
Storage Requirements Store at 4°C.
Regulatory Status RUO
Common Name LOC396892
Gene Symbol LOC396892
Assays Digestion of antibodies; Preparation of collagen for cosmeceutical purposes; Assessment of digestibility of proteins in food chemistry; Subculture of viable mammary epithelial cells.
Biological Activity ≥2,500 units per mg dry weight. One Unit releases 0.001 A280 as TCA soluble hydrolysis products from denatured hemoglobin per minute at 37°C.
Species Pig
Protein Tag None
Expression System Native protein purified from Porcine Stomach
Form Lyophilized
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.