Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Pepsin A Recombinant Rabbit Monoclonal Antibody (021), Invitrogen™
GREENER_CHOICE

Catalog No. PIMA543133
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543133 100 μL
PIMA543429 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543133 Supplier Invitrogen™ Supplier No. MA543133
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Specificity: Human PGA4

Pepsin consists of a single polypeptide chain and arises from its precursor, pepsinogen, by removal of a 41 amino acid segment from the N-terminus. Pepsinogen is synthesized in the stomach lining, and hydrochloric acid, also produced by the gastric mucosa, is necessary to convert the inactive enzyme and to maintain the optimum acidity (pH 1-3) for pepsin function. Pepsin is particularly effective in cleaving peptide bonds involving aromatic amino acids. Pepsin shows extremely broad specificity; although bonds involving phenylalanine and leucine are preferred, many others are also cleaved to some extent. Pepsin A is a member of the subfamily A1 within the pepsin family and is the predominant endopeptidase in the gastric juice of vertebrates. Pepsin A is inhibited by ovUS-1, a uterine serpin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pepsin A
Applications Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone 021
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene PGA3
Gene Accession No. P0DJD7, P0DJD8, P0DJD9
Gene Alias 1110035E17Rik; Pepf; pepsin A; pepsin A-3; Pepsin A-4; pepsin A-5; Pepsin F; pepsinogen 3, group I (pepsinogen A); pepsinogen 4, group I (pepsinogen A); pepsinogen 5, group I; pepsinogen 5, group I (pepsinogen A); pepsinogen A3; pepsinogen A4; pepsinogen A5; pepsinogen F; pepsinogen F protein; pepsinogen-3; Pepsinogen-4; pepsinogen-5; Pg5; PGA3; PGA4; PGA5
Gene Symbols PGA3, PGA4, PGA5
Host Species Rabbit
Immunogen Recombinant Human PGA4 Protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5222, 643834, 643847
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.